missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Apoptosis facilitator Bcl-2-like protein 14 Protein
A DNA sequence encoding the Homo sapiens (Human) Apoptosis facilitator Bcl-2-like protein 14, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8211-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_110393.1 | |
Apoptosis facilitator Bcl-2-like protein 14 Protein | |
Research Use Only | |
BCL2L14 | |
Recombinant Protein | |
E. coli | |
MCSTSGCDLE EIPLDDDDLN TIEFKILAYY TRHHVFKSTP ALFSPKLLRT RSLSQRGLGN CSANESWTEV SWPCRNSQSS EKAINLGKKK SSWKAFFGVV EKEDSQSTPA KVSAQGQRTL EYQDSHSQQW SRCLSNVEQC LEHEAVDPKV ISIANRVAEI VYSWPPPQAT QAGGFKSKEI FVTEGLSFQL QGHVPVASSS KKDEEEQILA KIVELLKYSG DQLERKLKKD KALMGHFQDG LSYSVFKTIT DQVLMGVDPR GESEVKAQGF KAALVIDVTA KLTAIDNHPM NRVLGFGTKY LKENFSPWIQ QHGGWEKILG ISHEEVD | |
Greater than 80% as determined by SDS-PAGE. |
79370 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |