missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Arginase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Arginase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5675-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000036.2 | |
Arginase 1 Protein | |
Research Use Only | |
ARG1 | |
Recombinant Protein | |
E. coli | |
MSAKSRTIGI IGAPFSKGQP RGGVEEGPTV LRKAGLLEKL KEQECDVKDY GDLPFADIPN DSPFQIVKNP RSVGKASEQL AGKVAEVKKN GRISLVLGGD HSLAIGSISG HARVHPDLGV IWVDAHTDIN TPLTTTSGNL HGQPVSFLLK ELKGKIPDVP GFSWVTPCIS AKDIVYIGLR DVDPGEHYIL KTLGIKYFSM TEVDRLGIGK VMEETLSYLL GRKKRPIHLS FDVDGLDPSF TPATGTPVVG GLTYREGLYI TEEIYKTGLL SGLDIMEVNP SLGKTPEEVT RTVNTAVAIT LACFGLAREG NHKPIDYLNP PK | |
Greater than 80% as determined by SDS-PAGE. |
383 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |