missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ART1 / CD296 / ARTC1 Protein
A DNA sequence encoding the Homo sapiens (Human) ART1 / CD296 / ARTC1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5681-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_004305.2 | |
ART1 / CD296 / ARTC1 Protein | |
Research Use Only | |
ART1 | |
Recombinant Protein | |
E. coli | |
QSHPITRRDL FSQEIQLDMA LASFDDQYAG CAAAMTAALP DLNHTEFQAN QVYADSWTLA SSQWQERQAR WPEWSLSPTR PSPPPLGFRD EHGVALLAYT ANSPLHKEFN AAVREAGRSR AHYLHHFSFK TLHFLLTEAL QLLGSGQRPP RCHQVFRGVH GLRFRPAGPR ATVRLGGFAS ASLKHVAAQQ FGEDTFFGIW TCLGAPIKGY SFFPGEEEVL IPPFETFQVI NASRLAQGPA RIYLRALGKH STYNCEYIKD KKCKSGPCHL DNS | |
Greater than 90% as determined by SDS-PAGE. |
417 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |