missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Arylsulfatase B Protein
A DNA sequence encoding the Homo sapiens (Human) Arylsulfatase B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5679-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000037.2 | |
Arylsulfatase B Protein | |
Research Use Only | |
ARSB | |
Recombinant Protein | |
E. coli | |
SGAGASRPPH LVFLLADDLG WNDVGFHGSR IRTPHLDALA AGGVLLDNYY TQPLCTPSRS QLLTGRYQIR TGLQHQIIWP CQPSCVPLDE KLLPQLLKEA GYTTHMVGKW HLGMYRKECL PTRRGFDTYF GYLLGSEDYY SHERCTLIDA LNVTRCALDF RDGEEVATGY KNMYSTNIFT KRAIALITNH PPEKPLFLYL ALQSVHEPLQ VPEEYLKPYD FIQDKNRHHY AGMVSLMDEA VGNVTAALKS SGLWNNTVFI FSTDNGGQTL AGGNNWPLRG RKWSLWEGGV RGVGFVASPL LKQKGVKNRE LIHISDWLPT LVKLARGHTN GTKPLDGFDV WKTISEGSPS PRIELLHNID PNFVDSSPCP RNSMAPAKDD SSLPEYSAFN TSVHAAIRHG NWKLLTGYPG CGYWFPPPSQ YNVSEIPSSD PPTKTLWLFD IDRDPEERHD LSREYPHIVT KLLSRLQFYH KHSVPVYFPA QDPRCDPKAT GVWGPWM | |
Greater than 90% as determined by SDS-PAGE. |
411 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |