missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATF7 Protein
A DNA sequence encoding the Homo sapiens (Human) ATF7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5689-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001123532.1 | |
ATF7 Protein | |
Research Use Only | |
ATF7 | |
Recombinant Protein | |
E. coli | |
MGDDRPFVCN ATGCGQRFTN EDHLAVHKHK HEMTLKFGPA RTDSVIIADQ TPTPTRFLKN CEEVGLFNEL ASSFEHEFKK AADEDEKKAR SRTVAKKLVV FRPRLFLLCF GIIFLIG | |
Greater than 80% as determined by SDS-PAGE. |
11016 | |
500 μg | |
Not determined. | |
Full Length of BC042363 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |