missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human B2M Protein
A cDNA sequence encoding the B2M was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11117-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
567 | |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human | |
MSRSVALAVL ALLSLSGLEA IQRTPKIQVY SRHPAENGKS NFLNCYVSGF HPSDIEVDLL KNGERIEKVE HSDLSFSKDW SFYLLYYTEF TPTEKDEYAC RVNHVTLSQP KIVKWDRDM | |
Greater than 98.0%. |
B2M Protein | |
Research Use Only | |
B2M | |
Human | |
Untagged | |
Lyophilized from 0.02M NH4HCO3. |