missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BIN2 Protein

A cDNA sequence encoding the BIN2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11163-1ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15906348

  • 137.00€ / 1µg
Spedizione stimata: 28-06-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Specifica

Specifica

BIN2 Protein
Research Use Only
Recombinant Protein
Baculovirus
ADPMRGMPGA RTSSSGASEN HRARGQGGGP QGVGRMAEGK AGGAAGLFAK QVQKKFSRAQ EKVLQKLGKA VETKDERFEQ SASNFYQQQA EGHKLYKDLK NFLSAVKVMH ESSKRVSETL QEIYSSEWDG HEELKAIVWN NDLLWEDYEE KLADQAVRTM EIYVAQFSEI KERIAKRGRK LVDYDSARHH LEAVQNAKKK DEAKTAKAEE EFNKAQTVFE DLNQELLEEL PILYNSRIGC YVTIFQNISN LRDVFYREMS KLNHNLYEVM SKLEKQHSNK VFVVKGLSSS SRRSLVISPP VRTATVSSPL TSPTSPSTLS LKSESESVSA TEDLAPDAAQ GEDNSEIKEL LEEEEIEKEG SEASSSEEDE PLPACNGPAQ AQPSPTTERA KSQEEVLPSS TTPSPGGALS PSGQPSSSAT EVVLRTRTAS EGSEQPKKRA SIQRTSAPPS RPPPPRATAS PRPSSGNIPS SPTASGGGSP TSPRASLGTG TASPRTSLEV SPNPEPPEKP VRTPEAKENE NIHNQNPEEL CTSPTLMTSQ VASEPGEAKK MEDKEKDNKL ISANSSEGQD QLQVSMVPEN NNLTAPEPQE EVSTSENPQL HHHHHH
Greater than 80.0% as determined by SDS-PAGE.
1 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
BIN2 protein solution (0.25 mg/ml) contains Phosphate buffered saline (pH 7.4), 20% glycerol, 1mM DTT and 0.1mM PMSF.
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali