missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bisphosphoglycerate mutase Protein
A DNA sequence encoding the Homo sapiens (Human) Bisphosphoglycerate mutase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5726-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001280014.1 | |
Bisphosphoglycerate mutase Protein | |
Research Use Only | |
BPGM | |
Recombinant Protein | |
E. coli | |
SKYKLIMLRH GEGAWNKENR FCSWVDQKLN SEGMEEARNC GKQLKALNFE FDLVFTSVLN RSIHTAWLIL EELGQEWVPV ESSWRLNERH YGALIGLNRE QMALNHGEEQ VRLWRRSYNV TPPPIEESHP YYQEIYNDRR YKVCDVPLDQ LPRSESLKDV LERLLPYWNE RIAPEVLRGK TILISAHGNS SRALLKHLEG ISDEDIINIT LPTGVPILLE LDENLRAVGP HQFLGDQEAI QAAIKKVEDQ GKVKQAKK | |
Greater than 80% as determined by SDS-PAGE. |
669 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |