missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BLyS / TNFSF13B / BAFF Protein
A DNA sequence encoding the Human BLyS / TNFSF13B / BAFF, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10357-ec-250ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001139117.1 | |
BLyS / TNFSF13B / BAFF Protein | |
Research Use Only | |
TNFSF13B | |
The ED50 as determined by a mouse splenocyte survival assay is 0.5-2 ug/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH7.4, with 0.02 % Tween-20, lyophilized |
10673 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MAVQGPEETV TQDCLQLIAD SETPTIQKGS YTFVPWLLSF KRGSALEEKE NKILVKETGY FFIYGQVLYT DKTYAMGHLI QRKKVHVFGD ELSLVTLFRC IQNMPETLPN NSCYSAGIAK LEEGDELQLA IPRENAQISL DGDVTFFGAL KLL | |
Greater than 95% as determined by SDS-PAGE and HPLC. |