missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bone sialoprotein 1
A DNA sequence encoding the Homo sapiens (Human) Bone sialoprotein 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6187-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_004958.2 | |
Bone sialoprotein 1 | |
Research Use Only | |
IBSP | |
Recombinant Protein | |
E. coli | |
AIQLPKKAGD ITNKATKEKE SDEEEEEEEE GNENEESEAE VDENEQGING TSTNSTEAEN GNGSSGGDNG EEGEEESVTG ANAEDTTETG RQGKGTSKTT TSPNGGFEPT TPPQVYRTTS PPFGKTTTVE YEGEYEYTGA NEYDNGYEIY ESE | |
Greater than 80% as determined by SDS-PAGE. |
3381 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |