missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BRD2 Protein

A cDNA sequence encoding the BRD2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11203-1mg

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15936468

  • 4090.00€ / 1mg
Spedizione stimata: 26-06-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Specifica

Specifica

BRD2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMLQNVTP HNKLPGEGNA GLLGLGPEAA APGKRIRKPS LLYEGFESPT MASVPALQLT PANPPPPEVS NPKKPGRVTN QLQYLHKVVM KALWKHQFAW PFRQPVDAVK LGLPDYHKII KQPMDMGTIK RRLENNYYWA ASECMQDFNT MFTNCYIYNK PTDDIVLMAQ TLEKIFLQKV ASMPQEEQEL VVTIPKNSHK KGAKLAALQG SVTSAHQVPA VSSVSHTALY TPPPEIPTTV LNIPHPSVIS SPLLKSLHSA GPPLLAVTAA PPAQPLAKKK GVKRKADTTT PTPTAILAPG SPASPPGSLE PKAARLPPMR RESGRPIKPP RKDLPDSQQQ HQSSKKGKLS EQLKHCNGIL KELLSKKHAA YAWPFYKPVD ASALGLHDYH DIIKHPMDLS TVKRKMENRD YRDAQEFAAD VRLMFSNCYK YNPPDHDVVA MARKLQDVFE FRYAKMPD
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
BRD2 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT.
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali