missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BTC / Betacellulin Protein
A DNA sequence encoding the Human BTC / Betacellulin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10416-ec-20ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001303892.1 | |
BTC / Betacellulin Protein | |
Research Use Only | |
BTC | |
The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 0.05 ng/ml, corresponding to a specific activity of > 2.0 M 107 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, with 0.02 % Tween-20, lyophilized |
685 | |
20 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
DGNSTRSPET NGLLCGDPEE NCAATTTQSK RKGHFSRCPK QYKHYCIKGR CRFVVAEQTP SCVCDEGYIG ARCERVDLFY | |
Greater than 98% as determined by SDS-PAGE and HPLC. |