missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BTN3A3 Protein
A DNA sequence encoding the Homo sapiens (Human) BTN3A3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5731-ye-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001229732.1 | |
BTN3A3 Protein | |
Research Use Only | |
BTN3A3 | |
Recombinant Protein | |
Yeast | |
QFSVLGPSGP ILAMVGEDAD LPCHLFPTMS AETMELRWVS SSLRQVVNVY ADGKEVEDRQ SAPYRGRTSI LRDGITAGKA ALRIHNVTAS DSGKYLCYFQ DGDFYEKALV ELKVAALGSD LHIEVKGYED GGIHLECRST GWYPQPQIKW SDTKGENIPA VEAPVVADGV GLYAVAASVI MRGSSGGGVS CIIRNSLLGL EKTASISIAD PFFRSAQPW | |
Greater than 90% as determined by SDS-PAGE. |
10384 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |