missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human c-MPL / CD110 / TPOR Protein
A DNA sequence encoding the Homo sapiens (Human) c-MPL / CD110 / TPOR, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6376-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_005364.1 | |
c-MPL / CD110 / TPOR Protein | |
Research Use Only | |
MPL | |
Recombinant Protein | |
E. coli | |
QDVSLLASDS EPLKCFSRTF EDLTCFWDEE EAAPSGTYQL LYAYPREKPR ACPLSSQSMP HFGTRYVCQF PDQEEVRLFF PLHLWVKNVF LNQTRTQRVL FVDSVGLPAP PSIIKAMGGS QPGELQISWE EPAPEISDFL RYELRYGPRD PKNSTGPTVI QLIATETCCP ALQRPHSASA LDQSPCAQPT MPWQDGPKQT SPSREASALT AEGGSCLISG LQPGNSYWLQ LRSEPDGISL GGSWGSWSLP VTVDLPGDAV ALGLQCFTLD LKNVTCQWQQ QDHASSQGFF YHSRARCCPR DRYPIWENCE EEEKTNPGLQ TPQFSRCHFK SRNDSIIHIL VEVTTAPGTV HSYLGSPFWI HQAVRLPTPN LHWREISSGH LELEWQHPSS WAAQETCYQL RYTGEGHQDW KVLEPPLGAR GGTLELRPRS RYRLQLRARL NGPTYQGPWS SWSDPTRVET ATETAW | |
Greater than 90% as determined by SDS-PAGE. |
4352 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |