missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C-type lectin domain family 4 member C Protein
A DNA sequence encoding the Homo sapiens (Human) C-type lectin domain family 4 member C, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7991-ma-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_569708.1 | |
C-type lectin domain family 4 member C Protein | |
Research Use Only | |
CLEC4C | |
Recombinant Protein | |
Mammal | |
NFMYSKTVKR LSKLREYQQY HPSLTCVMEG KDIEDWSCCP TPWTSFQSSC YFISTGMQSW TKSQKNCSVM GADLVVINTR EEQDFIIQNL KRNSSYFLGL SDPGGRRHWQ WVDQTPYNEN VTFWHSGEPN NLDERCAIIN FRSSEEWGWN DIHCHVPQKS ICKMKKIYI | |
Greater than 80% as determined by SDS-PAGE. |
170482 | |
100 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |