missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C-X-C chemokine receptor type 2 Protein
A DNA sequence encoding the Homo sapiens (Human) C-X-C chemokine receptor type 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6229-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001161770.1 | |
C-X-C chemokine receptor type 2 Protein | |
Research Use Only | |
CXCR2 | |
Recombinant Protein | |
E. coli | |
MEDFNMESDS FEDFWKGEDL SNYSYSSTLP PFLLDAAPCE | |
Greater than 90% as determined by SDS-PAGE. |
3579 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |