Learn More
enQuireBio™ Recombinant Human Casein kinase I isoform epsilon Protein
A DNA sequence encoding the Homo sapiens (Human) Casein kinase I isoform epsilon, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8358-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001276841.1 | |
Casein kinase I isoform epsilon Protein | |
Research Use Only | |
CSNK1E | |
Recombinant Protein | |
E. coli | |
SGSFGDIYLG ANIASGEEVA IKLECVKTKH PQLHIESKFY KMMQGGVGIP SIKWCGAEGD YNVMVMELLG PSLEDLFNFC SRKFSLKTVL LLADQMISRI EYIHSKNFIH RDVKPDNFLM GLGKKGNLVY IIDFGLAKKY RDARTHQHIP YRENKNLTGT ARYASINTHL GIEQSRRDDL ESLGYVLMYF NLGSLPWQGL KAATKRQKYE RISEK | |
Greater than 90% as determined by SDS-PAGE. |
102800317 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |