missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cathepsin F Protein
A DNA sequence encoding the Homo sapiens (Human) Cathepsin F, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8829-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_003784.2 | |
Cathepsin F Protein | |
Research Use Only | |
CTSF | |
Recombinant Protein | |
E. coli | |
PEWDWRSKGA VTKVKDQGMC GSCWAFSVTG NVEGQWFLNQ GTLLSLSEQE LLDCDKMDKA CMGGLPSNAY SAIKNLGGLE TEDDYSYQGH MQSCNFSAEK AKVYINDSVE LSQNEQKLAA WLAKRGPISV AINAFGMQFY RHGISRPLRP LCSPWLIDHA VLLVGYGNRS DVPFWAIKNS WGTDWGEKGY YYLHRGSGAC GVNTMASSAV VD | |
Greater than 90% as determined by SDS-PAGE. |
8722 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |