missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCDC101 Protein
A cDNA sequence encoding the CCDC101 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11286-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
CCDC101 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CCDC101 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM Nacl, 1mM DTT and 10% glycerol. |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MALVSADSRI AELLTELHQL IKQTQEERSR SEHNLVNIQK THERMQTENK ISPYYRTKLR GLYTTAKADA EAECNILRKA LDKIAEIKSL LEERRIAAKI AGLYNDSEPP RKTMRRGVLM TLLQQSAMTL PLWIGKPGDK PPPLCGAIPA SGDYVARPGD KVAARVKAVD GDEQWILAEV VSYSHATNKY EVDDIDEEGK ERHTLSRRRV IPLPQWKANP ETDPEALFQK EQLVLALYPQ TTCFYRALIH APPQRPQDDY SVLFEDTSYA DGYSPPLNVA QRYVVACKEP KKK |