missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL14 / HCC-1 / HCC-3 Protein
A DNA sequence encoding the Homo sapiens (Human) CCL14 / HCC-1 / HCC-3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7536-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_116738.1 | |
CCL14 / HCC-1 / HCC-3 Protein | |
Research Use Only | |
CCL14 | |
Recombinant Protein | |
E. coli | |
TKTESSSRGP YHPSECCFTY TTYKIPRQRI MDYYETNSQC SKPGIVFITK RGHSVCTNPS DKWVQDYIKD MKEN | |
Greater than 90% as determined by SDS-PAGE. |
6358 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |