Learn More
enQuireBio™ Recombinant Human CCL2 / MCP-1 / MCP1 Protein
A DNA sequence encoding the Human CCL2 / MCP-1 / MCP1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10224-ec-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002973.1 | |
CCL2 / MCP-1 / MCP1 Protein | |
Research Use Only | |
CCL2 | |
The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration range of 10-100 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
6347 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
QPDAINAPVT CCYNFTNRKI SVQRLASYRR ITSSKCPKEA VIFKTIVAKE ICADPKQKWV QDSMDHLDKQ TQTPKT | |
Greater than 96% as determined by SDS-PAGE and HPLC. |