missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCNG1 Protein
A cDNA sequence encoding the CCNG1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11296-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
900 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MIEVLTTTDS QKLLHQLNAL LEQESRCQPK VCGLRLIESA HDNGLRMTAR LRDFEVKDLL SLTQFFGFDT ETFSLAVNLL DRFLSKMKVQ PKHLGCVGLS CFYLAVKSIE EERNVPLATD LIRISQYRFT VSDLMRMEKI VLEKVCWKVK ATTAFQFLQL YYSLLQENLP LERRNSINFE RLEAQLKACH CRIIFSKAKP SVLALSIIAL EIQAQKCVEL TEGIECLQKH SKINGRDLTF WQELVSKCLT EYSSNKCSKP NVQKLKWIVS GRTARQLKHS YYRITHLPTI PEMVP | |
Greater than 85% as determined by SDS-PAGE. |
CCNG1 Protein | |
Research Use Only | |
CCNG1 | |
Human | |
His | |
CCNG1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 50% glycerol, 0.2M NaCl and 5mM DTT. |