missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD7 Protein
A DNA sequence encoding the Homo sapiens (Human) CD7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5794-ye-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_006128.1 | |
CD7 Protein | |
Research Use Only | |
CD7 | |
Recombinant Protein | |
Yeast | |
AQEVQQSPHC TTVPVGASVN ITCSTSGGLR GIYLRQLGPQ PQDIIYYEDG VVPTTDRRFR GRIDFSGSQD NLTITMHRLQ LSDTGTYTCQ AITEVNVYGS GTLVLVTEEQ SQGWHRCSDA PPRASALPAP PTGSALPDPQ TASALPDPPA ASALP | |
Greater than 90% as determined by SDS-PAGE. |
924 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |