missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD8A / MAL Protein
A DNA sequence encoding the Homo sapiens (Human) CD8A / MAL, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP2208-ma-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001139345.1 | |
CD8A / MAL Protein | |
100 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
925 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
CD8A | |
Recombinant Protein | |
Mammal | |
SQFRVSPLDR TWNLGETVEL KCQVLLSNPT SGCSWLFQPR GAAASPTFLL YLSQNKPKAA EGLDTQRFSG KRLGDTFVLT LSDFRRENEG YYFCSALSNS IMYFSHFVPV FLPAKPTTTP APRPPTPAPT IASQPLSLRP EACRPAAGGA VHTRGLDFAC D |