missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDKN2AIP N-terminal-like Protein
A DNA sequence encoding the Homo sapiens (Human) CDKN2AIP N-terminal-like protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5808-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_542387.1 | |
CDKN2AIP N-terminal-like Protein | |
Research Use Only | |
CDKN2AIPNL | |
Recombinant Protein | |
E. coli | |
MVGGEAAAAV EELVSGVRQA ADFAEQFRSY SESEKQWKAR MEFILRHLPD YRDPPDGSGR LDQLLSLSMV WANHLFLGCS YNKDLLDKVM EMADGIEVED LPQFTTRSEL MKKHQS | |
Greater than 90% as determined by SDS-PAGE. |
91368 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |