missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CEACAM4 Protein
A DNA sequence encoding the Homo sapiens (Human) CEACAM4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5812-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001808.2 | |
CEACAM4 Protein | |
Research Use Only | |
CEACAM4 | |
Recombinant Protein | |
Yeast | |
FTIEALPSSA AEGKDVLLLA CNISETIQAY YWHKGKTAEG SPLIAGYITD IQANIPGAAY SGRETVYPNG SLLFQNITLE DAGSYTLRTI NASYDSDQAT GQLHVHQNNV PGLPVGAVAG | |
Greater than 90% as determined by SDS-PAGE. |
1089 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |