missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Centrosomal protein of 63 kDa Protein
A DNA sequence encoding the Homo sapiens (Human) Centrosomal protein of 63 kDa, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7898-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001035842.1 | |
Centrosomal protein of 63 kDa Protein | |
Research Use Only | |
CEP63 | |
Recombinant Protein | |
E. coli | |
MEALLEGIQN RGHGGGFLTS CEAELQELMK QIDIMVAHKK SEWEGRTHAL ETCLKIREQE LKSLRSQLDV THKEVGMLHQ QVEEHEKIKQ EMTMEYKQEL KKLHEELCIL KRSYEKLQKK QMREFRGNTK NHREDRSEIE RLTAKIEEFR QKSLDWEKQR LIYQQQVSSL EAQRKALAEQ SEIIQAQLVN RKQKLESVEL SSQSEIQHLS SKLERANDTI CANELEIERL TMRVNDLVGT SMTVLQEQQQ KEEKLRESEK LLEALQEEKR ELKAALQSQE NLIHEARIQK EKLQEKVKAT NTQHAVEAIS LESVSATCKQ LSQELMEKYE ELKRMEAHNN EYKAEIKKLK EQILQGEQSY SSALEGMKME ISHLTQELHQ RDITIASTKG SSSDMEKRLR AEMQKAEDKA VEHKEILDQL ESLKLENRHL SEMVMKLELG LHECSLPVSP LGSIATRFLE EEELRSHHIL ERLDAHIEEL KRESEKTVRQ FTALK | |
Greater than 90% as determined by SDS-PAGE. |
80254 | |
1 mg | |
Not determined. | |
Full Length of Isoform 3 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |