missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CETN2 / Centrin 2 Protein
A DNA sequence encoding the Homo sapiens (Human) CETN2 / Centrin 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5820-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_004335.1 | |
CETN2 / Centrin 2 Protein | |
Research Use Only | |
CETN2 | |
Recombinant Protein | |
E. coli | |
MASNFKKANM ASSSQRKRMS PKPELTEEQK QEIREAFDLF DADGTGTIDV KELKVAMRAL GFEPKKEEIK KMISEIDKEG TGKMNFGDFL TVMTQKMSEK DTKEEILKAF KLFDDDETGK ISFKNLKRVA KELGENLTDE ELQEMIDEAD RDGDGEVSEQ EFLRIMKKTS LY | |
Greater than 90% as determined by SDS-PAGE. |
1069 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |