missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Charged multivesicular body protein 4b Protein
A DNA sequence encoding the Homo sapiens (Human) Charged multivesicular body protein 4b, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8215-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_789782.1 | |
Charged multivesicular body protein 4b Protein | |
Research Use Only | |
CHMP4B | |
Recombinant Protein | |
E. coli | |
SVFGKLFGAG GGKAGKGGPT PQEAIQRLRD TEEMLSKKQE FLEKKIEQEL TAAKKHGTKN KRAALQALKR KKRYEKQLAQ IDGTLSTIEF QREALENANT NTEVLKNMGY AAKAMKAAHD NMDIDKVDEL MQDIADQQEL AEEISTAISK PVGFGEEFDE DELMAELEEL EQEELDKNLL EISGPETVPL PNVPSIALPS KPAKKKEEED DDMKELENWA GSM | |
Greater than 80% as determined by SDS-PAGE. |
128866 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |