missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CHM Protein
A DNA sequence encoding the Homo sapiens (Human) CHM, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5830-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000381.1 | |
CHM Protein | |
Research Use Only | |
CHM | |
Recombinant Protein | |
E. coli | |
MADTLPSEFD VIVIGTGLPE SIIAAACSRS GRRVLHVDSR SYYGGNWASF SFSGLLSWLK EYQENSDIVS DSPVWQDQIL ENEEAIALSR KDKTIQHVEV FCYASQDLHE DVEEAGALQK NHALVTSANS TEAADSAFLP TEDESLSTMS CEMLTEQTPS SDPENALEVN GAEVTGEKEN HCDDKTCVPS TSAEDMSENV PIAEDTTEQP KKNRITYSQI IKEGRRFNID LVSKLLYSRG LLIDLLIKSN VSRYAEFKNI TRILAFREGR VEQVPCSRAD VFNSKQLTMV EKRMLMKFLT FCMEYEKYPD EYKGYEEITF YEYLKTQKLT PNLQYIVMHS IAMTSETASS TIDGLKATKN FLHCLGRYGN TPFLFPLYGQ GELPQCFCRM CAVFGGIYCL RHSVQCLVVD KESRKCKAII DQFGQRIISE HFLVEDSYFP ENMCSRVQYR QISRAVLITD RSVLKTDSDQ QISILTVPAE EPGTFAVRVI ELCSSTMTCM KGTYLVHLTC TSSKTAREDL ESVVQKLFVP YTEMEIENEQ VEKPRILWAL YFNMRDSSDI SRSCYNDLPS NVYVCSGPDC GLGNDNAVKQ AETLFQEICP NEDFCPPPPN PEDIILDGDS LQPEASESSA IPEANSETFK ESTNLGNLEE SSE | |
Greater than 90% as determined by SDS-PAGE. |
1121 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |