missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CIB1 Protein
A cDNA sequence encoding the CIB1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11428-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
10519 | |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
CIB1 | |
Human | |
His | |
The CIB1 protein solution (1 mg/ml) contains 20mM Tris-HCl, pH 8 and 5mM DTT. |
CIB1 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGGSGSRLSK ELLAEYQDLT FLTKQEILLA HRRFCELLPQ EQRTVESSLR AQVPFEQILS LPELKANPFK ERICRVFSTS PAKDSLSFED FLDLLSVFSD TATPDIKSHY AFRIFDFDDD GTLNREDLSR LVNCLTGEGE DTRLSASEMK QLIDNILEES DIDRDGTINL SEFQHVISRS PDFASSFKIV L |