missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CL-K1 / COLEC11 Protein
A DNA sequence encoding the Homo sapiens (Human) CL-K1 / COLEC11, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10143-ma-20ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001242911.1 | |
CL-K1 / COLEC11 Protein | |
Research Use Only | |
COLEC11 | |
Recombinant Protein | |
Mammal | |
QPAGDDACSV QILVPGLKGD AGEKGDKGAP GRPGRVGPTG EKGDMGDKGQ KGSVGRHGKI GPIGSKGEKG DSGDIGPPGP NGEPGLPCEC SQLRKAIGEM DNQVSQLTSE LKFIKNAVAG VRETESKIYL LVKEEKRYAD AQLSCQGRGG TLSMPKDEAA NGLMAAYLAQ AGLARVFIGI NDLEKEGAFV YSDHSPMRTF NKWRSGEPNN AYDEEDCVEM VASGGWNDVA CHTTMYFMCE FDKENM | |
Greater than 80% as determined by SDS-PAGE. |
78989 | |
20 μg | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
His | |
Tris-based buffer, 50% glycerol |