missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CLNS1A Protein
A cDNA sequence encoding the CLNS1A was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11450-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
CLNS1A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSFLKS FPPPGPAEGL LRQQPDTEAV LNGKGLGTGT LYIAESRLSW LDGSGLGFSL EYPTISLHAL SRDRSDCLGE HLYVMVNAKF EEESKEPVAD EEEEDSDDDV EPITEFRFVP SDKSALEAMF TAMCECQALH PDPEDEDSDD YDGEEYDVEA HEQGQGDIPT FYTYEEGLSH LTAEGQATLE RLEGMLSQSV SSQYNMAGVR TEDSIRDYED GMEVDTTPTV AGQFEDADVD H | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CLNS1A solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 2mM DTT. |