missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CPVL / Carboxypeptidase Protein
A DNA sequence encoding the Homo sapiens (Human) CPVL / Carboxypeptidase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8142-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001334981.1 | |
CPVL / Carboxypeptidase Protein | |
Research Use Only | |
CPVL | |
Recombinant Protein | |
E. coli | |
LFRSLYRSVS MPPKGDSGQP LFLTPYIEAG KIQKGRELSL VGPFPGLNMK SYAGFLTVNK TYNSNLFFWF FPAQIQPEDA PVVLWLQGGP GGSSMFGLFV EHGPYVVTSN MTLRDRDFPW TTTLSMLYID NPVGTGFSFT DDTHGYAVNE DDVARDLYSA LIQFFQIFPE YKNNDFYVTG ESYAGKYVPA IAHLIHSLNP VREVKINLNG IAIGDGYSDP ESIIGGYAEF LYQIGLLDEK QKKYFQKQCH ECIEHIRKQN WFEAFEILDK LLDGDLTSDP SYFQNVTGCS NYYNFLRCTE PEDQLYYVKF LSLPEVRQAI HVGNQTFNDG TIVEKYLRED TVQSVKPWLT EIMNNYKVLI YNGQLDIIVA AALTERSLMG MDWKGSQEYK KAEKKVWKIF KSDSEVAGYI RQAGDFHQVI IRGGGHILPY DQPLRAFDMI NRFIYGKGWD PYVG | |
Greater than 90% as determined by SDS-PAGE. |
54504 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |