missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CREB1 Protein
A DNA sequence encoding the Homo sapiens (Human) CREB1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5866-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_004370.1 | |
CREB1 Protein | |
Research Use Only | |
CREB1 | |
Recombinant Protein | |
E. coli | |
MTMESGAENQ QSGDAAVTEA ENQQMTVQAQ PQIATLAQVS MPAAHATSSA PTVTLVQLPN GQTVQVHGVI QAAQPSVIQS PQVQTVQSSC KDLKRLFSGT QISTIAESED SQESVDSVTD SQKRREILSR RPSYRKILND LSSDAPGVPR IEEEKSEEET SAPAITTVTV PTPIYQTSSG QYIAITQGGA IQLANNGTDG VQGLQTLTMT NAAATQPGTT ILQYAQTTDG QQILVPSNQV VVQAASGDVQ TYQIRTAPTS TIAPGVVMAS SPALPTQPAE EAARKREVRL MKNREAAREC RRKKKEYVKC LENRVAVLEN QNKTLIEELK ALKDLYCHKS D | |
Greater than 90% as determined by SDS-PAGE. |
1385 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |