missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CRIP2 Protein
A DNA sequence encoding the Homo sapiens (Human) CRIP2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5869-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001257766.1 | |
CRIP2 Protein | |
Research Use Only | |
CRIP2 | |
Recombinant Protein | |
E. coli | |
MASKCPKCDK TVYFAEKVSS LGKDWHKFCL KCERCSKTLT PGGHAEHDGK PFCHKPCYAT LFGPKGVNIG GAGSYIYEKP LAEGPQVTGP IEVPAARAEE RKASGPPKGP SRASSVTTFT GEPNTCPRCS KKVYFAEKVT SLGKDWHRPC LRCERCGKTL TPGGHAEHDG QPYCHKPCYG ILFGPKGVNT GAVGSYIYDR DPEGKVQP | |
Greater than 90% as determined by SDS-PAGE. |
1397 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |