missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CRISP3 Protein
A cDNA sequence encoding the CRISP3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11507-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
CRISP3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASNEDKDPAFTA LLTTQTQVQR EIVNKHNELR RAVSPPARNM LKMEWNKEAA ANAQKWANQC NYRHSNPKDR MTSLKCGENL YMSSASSSWS QAIQSWFDEY NDFDFGVGPK TPNAVVGHYT QVVWYSSYLV GCGNAYCPNQ KVLKYYYVCQ YCPAGNWANR LYVPYEQGAP CASCPDNCDD GLCTNGCKYE DLYSNCKSLK LTLTCKHQLV RDSCKASCNC SNSIY | |
Greater than 85.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CRISP3 filtered (0.4microm) solution at a concentration of 0.3 mg/ml in 50mM acetate buffer pH 4, 5mM DTT and 20% glycerol. |