missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CRP Protein
A cDNA sequence encoding the CRP was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11513-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
1401 | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. | |
Research Use Only | |
CRP | |
Human | |
Untagged | |
The sterile protein solution contains 20mM Tris (pH 7.5), 2mM CaCl2, 0.14M NaCl and 0.05% NaN3. |
CRP Protein | |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MEKLLCFLVL TSLSHAFGQT DMSRKAFVFP KESDTSYVSL KAPLTKPLKA FTVCLHFYTE LSSTRGYSIF SYATKRQDNE ILIFWSKDIG YSFTVGGSEI LFEVPEVTVA PVHICTSWES ASGIVEFWVD GKPRVRKSLK KGYTVGAEAS IILGQEQDSF GGNFEGSQSL VGDIGNVNMW DFVLSPDEIN TIYLGGPFSP NVLNWRALKY EVQGEVFTKP QLWP |