missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CXCL1 / GRO-alpha Protein
Recombinant Protein
Marca: enQuireBio™ QP1008-500 ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001502.1 | |
CXCL1 / GRO-alpha Protein | |
Research Use Only | |
CXCL1 | |
Recombinant Protein | |
E. coli | |
ASVATELRCQ CLQTLQGIHP KNIQSVNVKS PGPHCAQTEV IATLKNGRKA CLNPASPIVK KIIEKMLNSD KSN | |
Greater than 95% as determined by SDS-PAGE. |
2919 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Bioactivity confirmed by human neutrophil chemotaxis assay (significanty different from control at Human CXCL1 / GRO-alpha concentrations greater than 10 ng/ml). | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |