missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CXCL12 / SDF-1 Protein
Recombinant Protein
Marca: enQuireBio™ QP5280-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000600.1 | |
CXCL12 / SDF-1 Protein | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human CXCL12 / SDF-1 induces primary human T cell chemotaxis (significanty different from control at Human CXCL12 / SDF-1 concentrations greater than 5-20 ng/ml). | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
6387 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
CXCL12 | |
Recombinant Protein | |
E. coli | |
KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRF KM |