missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cystatin B / CSTB Protein
A DNA sequence encoding the Homo sapiens (Human) Cystatin B / CSTB, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8313-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000091.1 | |
Cystatin B / CSTB Protein | |
Research Use Only | |
CSTB | |
Recombinant Protein | |
E. coli | |
MMCGAPSATQ PATAETQHIA DQVRSQLEEK ENKKFPVFKA VSFKSQVVAG TNYFIKVHVG DEDFVHLRVF QSLPHENKPL TLSNYQTNKA KHDELTYF | |
Greater than 90% as determined by SDS-PAGE. |
1476 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |