missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Deoxycytidylate deaminase Protein
A DNA sequence encoding the Homo sapiens (Human) Deoxycytidylate deaminase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5917-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001012750.1 | |
Deoxycytidylate deaminase Protein | |
Research Use Only | |
DCTD | |
Recombinant Protein | |
E. coli | |
MSEVSCKKRD DYLEWPEYFM AVAFLSAQRS KDPNSQVGAC IVNSENKIVG IGYNGMPNGC SDDVLPWRRT AENKLDTKYP YVCHAELNAI MNKNLTDVKG CSMYVALFPC NECAKLIIQA GIKEVIFMSD KYHDSDEATA ARLLFNMAGV TFRKFIPKCS KIVIDFDSIN SRPSQKLQ | |
Greater than 80% as determined by SDS-PAGE. |
1635 | |
10 μg | |
Not determined. | |
Full Length of BC001286 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |