missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Desert hedgehog Protein
A DNA sequence encoding the Human Desert hedgehog protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10185-ec-25ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_066382.1 | |
Desert hedgehog Protein | |
Research Use Only | |
DHH | |
The ED50 as determined by its ability to induce alkaline phosphatase production by C3H10T1/2(CCL-226) cells is 15-45 ug/ml. | |
Human | |
Untagged | |
0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized |
50846 | |
25 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
IIGPGRGPVG RRRYARKQLV PLLYKQFVPG VPERTLGASG PAEGRVARGS ERFRDLVPNY NPDIIFKDEE NSGADRLMTE RCKERVNALA IAVMNMWPGV RLRVTEGWDE DGHHAQDSLH YEGRALDITT SDRDRNKYGL LARLAVEAGF DWVYYESRNH VHVSVKADNS LAVRAGG | |
Greater than 96% as determined by SDS-PAGE and HPLC. |