missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Desmoglein-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Desmoglein-3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5948-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001935.2 | |
Desmoglein-3 Protein | |
Research Use Only | |
Dsg3 | |
Recombinant Protein | |
E. coli | |
EWVKFAKPCR EGEDNSKRNP IAKITSDYQA TQKITYRISG VGIDQPPFGI FVVDKNTGDI NITAIVDREE TPSFLITCRA LNAQGLDVEK PLILTVKILD INDNPPVFSQ QIFMGEIEEN SASNSLVMIL NATDADEPNH LNSKIAFKIV SQEPAGTPMF LLSRNTGEVR TLTNSLDREQ ASSYRLVVSG ADKDGEGLST QCECNIKVKD VNDNFPMFRD SQYSARIEEN ILSSELLRFQ VTDLDEEYTD NWLAVYFFTS GNEGNWFEIQ TDPRTNEGIL KVVKALDYEQ LQSVKLSIAV KNKAEFHQSV ISRYRVQSTP VTIQVINVRE GIAFRPASKT FTVQKGISSK KLVDYILGTY QAIDEDTNKA ASNVKYVMGR NDGGYLMIDS KTAEIKFVKN MNRDSTFIVN KTITAEVLAI DEYTGKTSTG TVYVRVPDFN DNCPTAVLEK DAVCSSSPSV VVSARTLNNR YTGPYTFALE DQPVKLPAVW SITTLNATSA LLRAQEQIPP GVYHISLVLT DSQNNRCEMP RSLTLEVCQC DNRGICGTSY PTTSPGTRYG RPHSGR | |
Greater than 90% as determined by SDS-PAGE. |
1830 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |