missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Diamine acetyltransferase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Diamine acetyltransferase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8424-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002961.1 | |
Diamine acetyltransferase 1 Protein | |
Research Use Only | |
SAT1 | |
Recombinant Protein | |
E. coli | |
VIRPATAADC SDILRLIKEL AKYEYMEEQV ILTEKDLLED GFGEHPFYHC LVAEVPKEHW TPEGHSIVGF AMYYFTYDPW IGKLLYLEDF FVMSDYRGFG IGSEILKNLS QVAMRCRCSS MHFLVAEWNE PSINFYKRRG ASDLSSEEGW RLFKIDKEYL LKMATEE | |
Greater than 90% as determined by SDS-PAGE. |
6303 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |