missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Diphthamide biosynthesis protein 1
A DNA sequence encoding the Homo sapiens (Human) Diphthamide biosynthesis protein 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8021-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001034678.1 | |
Diphthamide biosynthesis protein 1 | |
Research Use Only | |
DPH2 | |
Recombinant Protein | |
E. coli | |
MESMFSSPAE AALQRETGVP GLLTPLPDLD GVYELERVAG FVRDLGCERV ALQFPDQLLG DAVAVAARLE ETTGSKMFIL GDTAYGSCCV DVLGAEQAGA QALIHFGPAC LSPPARPLPV AFVLRQRSVA LELCVKAFEA QNPDPKAPVV LLSEPACAHA LEALATLLRP RYLDLLVSSP AFPQPVGSLS PEPMPLERFG RRFPLAPGRR LEEYGAFYVG GSKASPDPDL DPDLSRLLLG WAPGQPFSSC CPDTGKTQDE GARAGRLRAR RRYLVERARD ARVVGLLAGT LGVAQHREAL AHLRNLTQAA GKRSYVLALG RPTPAKLANF PEVDVFVLLA CPLGALAPQL SGSFFQPILA PCELEAACNP AWPPPGLAPH LTHYADLLPG SPFHVALPPP ESELWETPDV SLITGDLRPP PAWKSSNDHG SLALTPRPQL ELAESSPAAS FLSSRSWQGL EPRLGQTPVT EAVSGRRGIA IAYEDEGSG | |
Greater than 90% as determined by SDS-PAGE. |
1802 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |