missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA-directed RNA polymerase III subunit RPC1 Protein
A DNA sequence encoding the Homo sapiens (Human) DNA-directed RNA polymerase III subunit RPC1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6511-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_008986.2 | |
DNA-directed RNA polymerase III subunit RPC1 Protein | |
Research Use Only | |
POLR3A | |
Recombinant Protein | |
E. coli | |
FPEKVNKANI NFLRKLVQNG PEVHPGANFI QQRHTQMKRF LKYGNREKMA QELKYGDIVE RHLIDGDVVL FNRQPSLHKL SIMAHLARVK PHRTFRFNEC VCTPYNADFD GDEMNLHLPQ TEEAKAEALV LMGTKANLVT PRNGEPLIAA IQDFLTGAYL LTLKDTFFDR AKACQIIASI LVGKDEKIKV RLPPPTILKP VTLWTGKQIF SVILRPSDDN PVRANLRTKG KQYCGKGEDL C | |
Greater than 90% as determined by SDS-PAGE. |
11128 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |