missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA primase small subunit Protein
A DNA sequence encoding the Homo sapiens (Human) DNA primase small subunit, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6528-ec-1mg
Weitere Details : Peso : 0.01000kg
Spezifikation
NP_000937.1 | |
DNA primase small subunit Protein | |
Research Use Only | |
PRIM1 | |
Recombinant Protein | |
E. coli | |
METFDPTELP ELLKLYYRRL FPYSQYYRWL NYGGVIKNYF QHREFSFTLK DDIYIRYQSF NNQSDLEKEM QKMNPYKIDI GAVYSHRPNQ HNTVKLGAFQ AQEKELVFDI DMTDYDDVRR CCSSADICPK CWTLMTMAIR IIDRALKEDF GFKHRLWVYS GRRGVHCWVC DESVRKLSSA VRSGIVEYLS LVKGGQDVKK KVHLSEKIHP FIRKSINIIK KYFEEYALVN QDILENKESW DKILALVPET IHDELQQSFQ KSHNSLQRWE HLKKVASRYQ NNIKNDKYGP WLEWEIMLQY CFPRLDINVS KGINHLLKSP FSVHPKTGRI SVPIDLQKVD QFDPFTVPTI SFICRELDAI STNEEEKEEN EAESDVKHRT RDYKKTSLAP YVKVFEHFLE NLDKSRKGEL LKKSDLQKDF | |
Greater than 90% as determined by SDS-PAGE. |
5557 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |