missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DnaJ homolog subfamily B member 8 Protein
A DNA sequence encoding the Homo sapiens (Human) DnaJ homolog subfamily B member 8, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7884-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_699161.1 | |
DnaJ homolog subfamily B member 8 Protein | |
Research Use Only | |
DNAJB8 | |
Recombinant Protein | |
E. coli | |
MANYYEVLGV QASASPEDIK KAYRKLALRW HPDKNPDNKE EAEKKFKLVS EAYEVLSDSK KRSLYDRAGC DSWRAGGGAS TPYHSPFDTG YTFRNPEDIF REFFGGLDPF SFEFWDSPFN SDRGGRGHGL RGAFSAGFGE FPAFMEAFSS FNMLGCSGGS HTTFSSTSFG GSSSGSSGFK SVMSSTEMIN GHKVTTKRIV ENGQERVEVE EDGQLKSVTV NGKEQLKWMD SK | |
Greater than 90% as determined by SDS-PAGE. |
165721 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |