missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNAL4 Protein
A cDNA sequence encoding the DNAL4 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11690-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifications
DNAL4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGETEGKKDE ADYKRLQTFP LVRHSDMPEE MRVETMELCV TACEKFSNNN ESAAKMIKET MDKKFGSSWH VVIGEGFGFE ITHEVKNLLY LYFGGTLAVC VWKCS | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
DNAL4 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT. |